Learn More
Thermo Scientific Filament Assembly
Catalog No. NC9383699 Shop All Thermo Scientific Products
Encompass
A filament guide device used in fused filament fabrication 3D printers to guide and consistently feed filament into the extruder for material deposition.
Catalog No. NC9383699 Supplier Thermo Scientific Supplier No. 1027920
A filament guide device used in fused filament fabrication 3D printers to guide and consistently feed filament into the extruder for material deposition.