Learn More
Medchemexpress LLC Filanesib | 885060-09-3 | C20H22F2N4O2S | 100 MG
SDP
Catalog No. 5000372290
Encompass_Preferred
Filanesib is a selective and noncompetitive kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM for human KSP. It induces cell death by apoptosis in vitro and has potent anti-proliferative activity.
  • Induces mitotic arrest in multiple cell lines.
  • Exhibits anti-proliferative activity against a broad range of human and rodent tumor cell lines.
  • Induces apoptosis in a dose-dependent manner in HeLa cells.
  • Causes accumulation of cells in the G2/M phase of the cell cycle in HeLa cells.
  • Potently induces cell cycle block and subsequent death in leukemic cells via the mitochondrial pathway.
  • Able to induce caspase-2 activation.
  • Cytotoxic in Type II EOC cells.
  • Demonstrates anti-tumor activity in vivo.
Catalog No. 50-003-72290 Supplier Medchemexpress LLC Supplier No. HY15187100MG
Add to Cart
Add to Cart
Filanesib is a selective and noncompetitive kinesin spindle protein (KSP) inhibitor with an IC50 of 6 nM for human KSP. It induces cell death by apoptosis in vitro and has potent anti-proliferative activity.
  • Induces mitotic arrest in multiple cell lines.
  • Exhibits anti-proliferative activity against a broad range of human and rodent tumor cell lines.
  • Induces apoptosis in a dose-dependent manner in HeLa cells.
  • Causes accumulation of cells in the G2/M phase of the cell cycle in HeLa cells.
  • Potently induces cell cycle block and subsequent death in leukemic cells via the mitochondrial pathway.
  • Able to induce caspase-2 activation.
  • Cytotoxic in Type II EOC cells.
  • Demonstrates anti-tumor activity in vivo.