Learn More
Selleck Chemical LLC Filanesib hydrochloride
SDP
Catalog No. 5000004674
Encompass_Preferred
Filanesib (ARRY-520) hydrochloride is a kinesin spindle protein (KSP) inhibitor that blocks the separation of centrosomes and assembly of spindles in the early stage of cell mitosis and cell proliferation
Catalog No. 50-000-04674 Supplier Selleck Chemical LLC Supplier No. S73545MG
Add to Cart
Add to Cart
Filanesib (ARRY-520) hydrochloride is a kinesin spindle protein (KSP) inhibitor that blocks the separation of centrosomes and assembly of spindles in the early stage of cell mitosis and cell proliferation