Learn More
Medchemexpress LLC Filastatin | 431996-53-1 | MFCD03058358 | 99.3% | 359.8 g/mol | C18H18ClN3O3 | 50MG
SDP
Catalog No. 5000236174
Encompass_Preferred
Filastatin is a small-molecule antifungal research reagent that inhibits Candida albicans filamentation and adhesion. It blocks the yeast-to-hyphal morphological transition and inhibits adhesion across multiple pathogenic Candida species (IC50 ≈ 3 μM in a GFP-based adhesion assay), making it suitable for in vitro studies of fungal morphogenesis and host-pathogen interactions.
  • Inhibits Candida albicans filamentation and adhesion.
  • Reduces fungal adhesion to surfaces and human cells.
  • Blocks the yeast-to-hyphal morphological transition.
  • Useful for in vitro antifungal and pathogenesis research.
  • High purity appropriate for biochemical assays.
Catalog No. 50-002-36174 Supplier Medchemexpress LLC Supplier No. HY12470150MG
Filastatin is a small-molecule antifungal research reagent that inhibits Candida albicans filamentation and adhesion. It blocks the yeast-to-hyphal morphological transition and inhibits adhesion across multiple pathogenic Candida species (IC50 ≈ 3 μM in a GFP-based adhesion assay), making it suitable for in vitro studies of fungal morphogenesis and host-pathogen interactions.
  • Inhibits Candida albicans filamentation and adhesion.
  • Reduces fungal adhesion to surfaces and human cells.
  • Blocks the yeast-to-hyphal morphological transition.
  • Useful for in vitro antifungal and pathogenesis research.
  • High purity appropriate for biochemical assays.