Learn More
Cayman Chemical FIlgotInIb-d4 1mg
SDP
Supplier:  Cayman Chemical 288751 MG
Encompass_Preferred
An internal standard for the quantification of filgotinib by GC- or LC-MS
Catalog No. 50-293-8455