Learn More
Selleck Chemical LLC Filgotinib (GLPG0634) 25mg 1206161-97-8
SDP
Supplier:  Selleck Chemical LLC S760525MG
Encompass_Preferred
Filgotinib (GLPG0634) is a selective JAK1 inhibitor with IC50 of 10 nM, 28 nM, 810 nM, and 116 nM for JAK1, JAK2, JAK3, and TYK2, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5009