Learn More
Medchemexpress LLC Filgotinib (maleate) | 1802998-75-9 | 50 MG
SDP
Catalog No. 5000387573
Encompass_Preferred
Filgotinib maleate (GLPG0634 maleate) is a selective, orally available JAK1 inhibitor with anti-inflammatory and antiviral activities. It inhibits JAK1, JAK2, JAK3, and TYK2 with varying IC50 values. This compound also demonstrates the ability to inhibit HIV-1 driven gene transcription and reduce the proliferation of HIV-1 infected cells, making it suitable for studies related to rheumatoid arthritis and inflammatory bowel disease.
  • Selectively inhibits JAK1
  • Possesses anti-inflammatory and antiviral properties
  • Inhibits HIV-1 driven gene transcription
  • Reduces proliferation of HIV-1 infected cells
  • Applicable for rheumatoid arthritis research
  • Applicable for inflammatory bowel disease research
Catalog No. 50-003-87573 Supplier Medchemexpress LLC Supplier No. HY18300A50MG
Filgotinib maleate (GLPG0634 maleate) is a selective, orally available JAK1 inhibitor with anti-inflammatory and antiviral activities. It inhibits JAK1, JAK2, JAK3, and TYK2 with varying IC50 values. This compound also demonstrates the ability to inhibit HIV-1 driven gene transcription and reduce the proliferation of HIV-1 infected cells, making it suitable for studies related to rheumatoid arthritis and inflammatory bowel disease.
  • Selectively inhibits JAK1
  • Possesses anti-inflammatory and antiviral properties
  • Inhibits HIV-1 driven gene transcription
  • Reduces proliferation of HIV-1 infected cells
  • Applicable for rheumatoid arthritis research
  • Applicable for inflammatory bowel disease research