Learn More
Medchemexpress LLC Filgotinib (maleate) | 1802998-75-9 | 99.88% | 541.58 | 1 ML
SDP
Catalog No. 5000387637
Encompass_Preferred
Filgotinib maleate (GLPG0634 maleate) is a selective, orally available JAK1 inhibitor with anti-inflammatory and antiviral activities. It can effectively inhibit the activities of JAK1, JAK2, JAK3, and TYK2. This compound also inhibits HIV-1 driven gene transcription and reduces proliferation of HIV-1 infected cells. It can be used in the study of rheumatoid arthritis and inflammatory bowel disease.
  • Selective, orally available JAK1 inhibitor
  • Anti-inflammatory and antiviral activities
  • Inhibits activities of JAK1, JAK2, JAK3, and TYK2
  • Inhibits HIV-1 driven gene transcription
  • Reduces proliferation of HIV-1 infected cells
  • Suitable for research in rheumatoid arthritis and inflammatory bowel disease
Catalog No. 50-003-87637 Supplier Medchemexpress LLC Supplier No. HY18300A10MM/1ML
Filgotinib maleate (GLPG0634 maleate) is a selective, orally available JAK1 inhibitor with anti-inflammatory and antiviral activities. It can effectively inhibit the activities of JAK1, JAK2, JAK3, and TYK2. This compound also inhibits HIV-1 driven gene transcription and reduces proliferation of HIV-1 infected cells. It can be used in the study of rheumatoid arthritis and inflammatory bowel disease.
  • Selective, orally available JAK1 inhibitor
  • Anti-inflammatory and antiviral activities
  • Inhibits activities of JAK1, JAK2, JAK3, and TYK2
  • Inhibits HIV-1 driven gene transcription
  • Reduces proliferation of HIV-1 infected cells
  • Suitable for research in rheumatoid arthritis and inflammatory bowel disease