Learn More
Medchemexpress LLC Filgotinib (maleate) | 1802998-75-9 | 99.9% | 541.58 | 5 MG
SDP
Catalog No. 5000387679
Encompass_Preferred
Filgotinib maleate is a selective, orally available JAK1 inhibitor with anti-inflammatory and antiviral activities. It effectively inhibits JAK1, JAK2, JAK3, and TYK2. It also inhibits HIV-1 driven gene transcription and reduces proliferation of HIV-1 infected cells. It can be used in the study of rheumatoid arthritis and inflammatory bowel disease.
  • Inhibits differentiation of Th2 cells and Th1 cells.
  • Produces dose-dependent bone damage protection.
  • Prevents the development of inflammation in animal models of collagen-induced arthritis.
  • Reduces inflammatory cell infiltration.
  • Protects articular cartilage and bone.
  • Reduces serum levels of inflammatory cytokines and chemokines.
Catalog No. 50-003-87679 Supplier Medchemexpress LLC Supplier No. HY18300A5MG
Filgotinib maleate is a selective, orally available JAK1 inhibitor with anti-inflammatory and antiviral activities. It effectively inhibits JAK1, JAK2, JAK3, and TYK2. It also inhibits HIV-1 driven gene transcription and reduces proliferation of HIV-1 infected cells. It can be used in the study of rheumatoid arthritis and inflammatory bowel disease.
  • Inhibits differentiation of Th2 cells and Th1 cells.
  • Produces dose-dependent bone damage protection.
  • Prevents the development of inflammation in animal models of collagen-induced arthritis.
  • Reduces inflammatory cell infiltration.
  • Protects articular cartilage and bone.
  • Reduces serum levels of inflammatory cytokines and chemokines.