Learn More
Medchemexpress LLC Filgotinib maleate | 1802998-75-9 | MFCD34184747 | 99.9% | 541.58 | C25H27N5O7S | 10 MG
SDP
Catalog No. 5000294877
Encompass_Preferred
Filgotinib maleate is the maleate salt of filgotinib, a selective, orally active JAK1 inhibitor supplied as a high-purity research reagent for studying inflammatory and antiviral pathways. Provided as a 10 mg solid, it is intended for laboratory use with recommended storage conditions noted on the product datasheet and SDS.
  • Selective JAK1 inhibitor for research applications.
  • Maleate salt form for improved solubility.
  • High purity suitable for analytical and biological studies.
  • 10 mg solid package convenient for small-scale experiments.
  • Recommended storage: solid 4°C; in solvent -80°C up to 6 months.
Catalog No. 50-002-94877 Supplier Medchemexpress LLC Supplier No. HY18300A10MG
Filgotinib maleate is the maleate salt of filgotinib, a selective, orally active JAK1 inhibitor supplied as a high-purity research reagent for studying inflammatory and antiviral pathways. Provided as a 10 mg solid, it is intended for laboratory use with recommended storage conditions noted on the product datasheet and SDS.
  • Selective JAK1 inhibitor for research applications.
  • Maleate salt form for improved solubility.
  • High purity suitable for analytical and biological studies.
  • 10 mg solid package convenient for small-scale experiments.
  • Recommended storage: solid 4°C; in solvent -80°C up to 6 months.