Learn More
TARGETMOL CHEMICALS INC FILIBUVIR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T152825MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Filibuvir (PF-00868554) is a selective and noncovalent inhibitor of HCV NS5B RNA-dependent RNA polymerase. Filibuvir inhibits genotype 1a and 1b replicons with EC50s of 59 nM. purity: 99%
Catalog No. 50-258-1234