Learn More
Cayman Chemical Filipin Complex, Purity: ≥95%, CAS: 11078-21-0, 50 mg
SDP
Catalog No. 502934469
Encompass_Preferred
A neutral polyene with antifungal activity; inhibits cell growth as well as mitochondrial terminal electron transport in S. cerevisiae at 135 µg/ml; binds to various sterols in aqueous solutions and fungal cell membranes at 50 µg/ml; induces membrane pit formation and leakage of cell contents; has been used to label sterols within biological structures for imaging; ex/em = 338/480 nm, respectively
Catalog No. 50-293-4469 Supplier Cayman Chemical Supplier No. 2507350 MG
A neutral polyene with antifungal activity; inhibits cell growth as well as mitochondrial terminal electron transport in S. cerevisiae at 135 µg/ml; binds to various sterols in aqueous solutions and fungal cell membranes at 50 µg/ml; induces membrane pit formation and leakage of cell contents; has been used to label sterols within biological structures for imaging; ex/em = 338/480 nm, respectively