Learn More
Medchemexpress LLC HY-N6716 5mg Medchemexpress, Filipin complex CAS:11078-21-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67165MG
Encompass_Preferred
Medchemexpress, HY-N6716 5mg Filipin complex CAS:11078-21-0 Filipin, produced as a mixture of related compounds known as the filipin complex (filipins I-IV) in nature, is a 28-membered ring pentaene macrolide antifungal antibiotic produced by S. filipinensis, S. avermitilis and S. miharaensis. Filipin interacts with membrane sterols causing the alteration of membrane structure. Filipin III is the major component of Filipin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0459