Learn More
Sigma Aldrich Fine Chemicals Biosciences filipin complex ready made
Catalog No. 502905042
Encompass
filipin Complex the collective name given to family of isomeric polyene macrolides isolated from cultures of S. filipinensis with filipin III being the majority part of the complex.
Catalog No. 50-290-5042 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE00881ML
filipin Complex the collective name given to family of isomeric polyene macrolides isolated from cultures of S. filipinensis with filipin III being the majority part of the complex.