Learn More
Cayman Chemical Filipin III, 480-49-9, 1 mg
SDP
Supplier:  Cayman Chemical 704401
Encompass_Preferred
Molecular Formula C35H58O11, Purity ≥90%, Formula Weight 654.8; A fluorescent molecule that can be used to detect cholesterol by fluorescence and freeze-fracture electron microscopy.
Catalog No. NC9384264