Learn More
Selleck Chemical LLC Filipin III
SDP
Supplier:  Selleck Chemical LLC S74021MG
Encompass_Preferred
Filipin III is a polyene macrolide antibiotic derived from Streptomyces filipinensis It exhibits antifungal activity against Candida utilis with an MIC of 0 3 mg/L and Saccharomyces cerevisiae with an MIC of 0 4 mg/L by interacting with membrane sterols leading to alterations in membrane structure Filipin III has a strong affinity for cholesterol and exhibits fluorescent properties with the potential to be used as a valuable tool for detecting and quantifying cholesterol in biological membranes
Catalog No. 50-000-01301