Learn More
Medchemexpress LLC HY-N6718 1mg Medchemexpress, Filipin III CAS:480-49-9 Purity:>98%
SDP
Catalog No. 501933969
Encompass_Preferred
Medchemexpress, HY-N6718 1mg Filipin III CAS:480-49-9 Filipin III is the major component of Filipin, a 28-membered ring pentaene macrolide antifungal antibiotic produced by S. filipinensis, S. avermitilis and S. miharaensis. Filipin interacts with membrane sterols causing the alteration of membrane structure [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3969 Supplier Medchemexpress LLC Supplier No. HYN67181MG
Medchemexpress, HY-N6718 1mg Filipin III CAS:480-49-9 Filipin III is the major component of Filipin, a 28-membered ring pentaene macrolide antifungal antibiotic produced by S. filipinensis, S. avermitilis and S. miharaensis. Filipin interacts with membrane sterols causing the alteration of membrane structure [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.