Learn More
Sigma Aldrich Fine Chemicals Biosciences Filipin III from Streptomyces filipinensis, 480-49-9, MFCD00466923, 1 mg
Catalog No. NC0215088
Encompass
Empirical Formula (Hill Notation): C35H58O11, Molecular Weight: 654.83, ≥85% (HPLC). Filipin has been used in a double staining procedure as a probe for the detection of lipoproteins in polyacrylamide gel and immobilized on nitrocellulose membranes. It is also widely used to localize and quantitate unesterified cholesterol by virtue of a specific fluorescent complex.
Catalog No. NC0215088 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F47671MG
Empirical Formula (Hill Notation): C35H58O11, Molecular Weight: 654.83, ≥85% (HPLC). Filipin has been used in a double staining procedure as a probe for the detection of lipoproteins in polyacrylamide gel and immobilized on nitrocellulose membranes. It is also widely used to localize and quantitate unesterified cholesterol by virtue of a specific fluorescent complex.