Learn More
Cole-Parmer Oakton Electrode Fill Solution, 3.8M KCL, 2/pack
Catalog No. 50633681 Shop All Cole-Parmer Products
Encompass_Preferred
This solution comes in a pack of two 125 mL bottles, which is enough to fill about 25 electrodes.
Catalog No. 50-633-681 Supplier Cole-Parmer Supplier No. 0599247
This solution comes in a pack of two 125 mL bottles, which is enough to fill about 25 electrodes.