Learn More
Grainger Filler Maple 192 oz
Supplier:  Grainger 16NH06
Encompass_Preferred
FAMOWOOD Filler Original Wood Filler is a wood filler that can be stained, sanded, and drilled. It is a solvent-based wood filler that dries fast and hard. It can fill cracks and holes in wood up to 1 inch deep. It is ready to sand in 15 minutes and ready to paint or stain in 1 hour. It comes in a 16 oz tub. The filler is designed for interior and exterior wood repairs. It can be used on hardwoods, plywood, paneling, furniture, cabinets, windows, doors, trim, and more. It will not shrink, crack or fall out. It cleans up easily with mineral spirits.
Catalog No. 50-276-4535