Learn More
Grainger Filler Red Wood 192 oz
Catalog No. 502764598
Encompass_Preferred
The FAMOWOOD 16NG90 Glaze Coat is a clear, water-based polyurethane finish that provides a durable protective coating for wood surfaces. It dries to a crystal clear finish, enhancing the natural beauty of wood grain. This 32 oz can requires no stirring or thinning and cleans up easily with soap and water. It dries tack-free in 2 hours and covers about 150 sq ft per coat. Ideal for interior wood surfaces like furniture, cabinets, doors, and trim, it offers excellent abrasion resistance and protection from household chemicals. Non-flammable when wet, it resists chemicals, heat, and water once cured. Available online from Grainger.
Catalog No. 50-276-4598 Supplier Grainger Supplier No. 16NG90
The FAMOWOOD 16NG90 Glaze Coat is a clear, water-based polyurethane finish that provides a durable protective coating for wood surfaces. It dries to a crystal clear finish, enhancing the natural beauty of wood grain. This 32 oz can requires no stirring or thinning and cleans up easily with soap and water. It dries tack-free in 2 hours and covers about 150 sq ft per coat. Ideal for interior wood surfaces like furniture, cabinets, doors, and trim, it offers excellent abrasion resistance and protection from household chemicals. Non-flammable when wet, it resists chemicals, heat, and water once cured. Available online from Grainger.