Learn More
Grainger Filler Tan 192 oz
Catalog No. 502764597
Encompass_Preferred
FAMOWOOD Filler Original Wood Filler is a water-based wood filler that can be stained, sanded and drilled. It is designed for repairing defects in wood surfaces and can fill cracks up to 1 inch wide. The filler dries quickly in 15-20 minutes and can be stained or painted in 1 hour. It comes in a 16 oz plastic tub. The filler is easy to apply, dries to a natural wood color and can be used for interior and exterior wood repairs. It sands easily and cleans up with water before drying.
Catalog No. 50-276-4597 Supplier Grainger Supplier No. 16NG87
FAMOWOOD Filler Original Wood Filler is a water-based wood filler that can be stained, sanded and drilled. It is designed for repairing defects in wood surfaces and can fill cracks up to 1 inch wide. The filler dries quickly in 15-20 minutes and can be stained or painted in 1 hour. It comes in a 16 oz plastic tub. The filler is easy to apply, dries to a natural wood color and can be used for interior and exterior wood repairs. It sands easily and cleans up with water before drying.