Learn More
Grainger Filler Tan 192 oz
Catalog No. 502763742
Encompass_Preferred
FAMOWOOD Filler Original Wood Filler is a wood filler that can be used to fill cracks, nail holes, and other defects in wood. It is a water-based, acrylic latex formula that dries quickly and can be stained, sanded, drilled, screwed or planed. It cleans up easily with water and is non-toxic when dry. The filler comes in a 16 oz plastic tub and is available in natural and various wood tone colors to blend with different types of wood. It bonds tightly to wood and can be used for interior and exterior applications. The filler will not shrink, crack or fall out.
Catalog No. 50-276-3742 Supplier Grainger Supplier No. 16NH08
FAMOWOOD Filler Original Wood Filler is a wood filler that can be used to fill cracks, nail holes, and other defects in wood. It is a water-based, acrylic latex formula that dries quickly and can be stained, sanded, drilled, screwed or planed. It cleans up easily with water and is non-toxic when dry. The filler comes in a 16 oz plastic tub and is available in natural and various wood tone colors to blend with different types of wood. It bonds tightly to wood and can be used for interior and exterior applications. The filler will not shrink, crack or fall out.