Learn More
Grainger Filler White Pine 192 oz
Catalog No. 502764583
Encompass_Preferred
DAP Filler Painter's Putty is a high-performance, lightweight spackling for repairing holes, cracks and defects on interior walls and ceilings. It features an ultra-smooth formula that dries fast and sands easily without shrinking. This spackling compound spreads smoothly without sagging and one application fills holes up to 1 inch deep. It can be painted over in 30 minutes with latex or oil-based paints. This 1 quart tub of DAP spackling is designed for hand application using a putty knife or spatula. It's ideal for drywall repairs and small patching jobs.
Catalog No. 50-276-4583 Supplier Grainger Supplier No. 16NG89
DAP Filler Painter's Putty is a high-performance, lightweight spackling for repairing holes, cracks and defects on interior walls and ceilings. It features an ultra-smooth formula that dries fast and sands easily without shrinking. This spackling compound spreads smoothly without sagging and one application fills holes up to 1 inch deep. It can be painted over in 30 minutes with latex or oil-based paints. This 1 quart tub of DAP spackling is designed for hand application using a putty knife or spatula. It's ideal for drywall repairs and small patching jobs.