Learn More
Hach Company Filling Solution KCl 100mL
Catalog No. 5000207066 Shop All Hach Company Products
Encompass
Filling Solution-Reference-Saturated KCl (KCl L)-100 mLFilling solution for electrode reference element Potassium Chloride (KCl) 100 mL bottle
Catalog No. 50-002-07066 Supplier Hach Company Supplier No. S21M002
Filling Solution-Reference-Saturated KCl (KCl L)-100 mLFilling solution for electrode reference element Potassium Chloride (KCl) 100 mL bottle