Learn More
Anton Paar 600mm elongated filling tube for tanks and drums
Catalog No. NC1818218
Encompass_Preferred
Sometimes samples are hard to reach with the standard filling tube, e.g. when tanks or drums are not fully filled. In such cases you can remove the standard filling tube and use the elongated filling tube for more convenient sample filling. Suitable for use with Anton Paar density metersDMA 35 Standard Advanced portable density and concentration meter NC1488853,Snap 41 Portable Alcohol Meter for sugar-free distilled spirits NC1818207
Catalog No. NC1818218 Supplier Anton Paar Supplier No. 178843
Sometimes samples are hard to reach with the standard filling tube, e.g. when tanks or drums are not fully filled. In such cases you can remove the standard filling tube and use the elongated filling tube for more convenient sample filling. Suitable for use with Anton Paar density metersDMA 35 Standard Advanced portable density and concentration meter NC1488853,Snap 41 Portable Alcohol Meter for sugar-free distilled spirits NC1818207