Learn More
Alzet ALZETTM Filling Tube - Designed for Filling ALZET Osmotic Pumps of 100 µl Size
SDP
Catalog No. NC0837218
Encompass
These ALZETTM Filling Tubes are customized needles designed for filling any model of 100 µl size ALZET Osmotic Pump. •Length is customized to fit 100 µl ALZET pumps•Blunt-tipped to avoid damage to the pump reservoir•Luer lock for secure attachment to a syringe•Sterile and ready to useNote: ALZET pumps should only be filled with these specially designed filling tubes
Catalog No. NC0837218 Supplier Alzet Supplier No. 0007988PK
These ALZETTM Filling Tubes are customized needles designed for filling any model of 100 µl size ALZET Osmotic Pump. •Length is customized to fit 100 µl ALZET pumps•Blunt-tipped to avoid damage to the pump reservoir•Luer lock for secure attachment to a syringe•Sterile and ready to useNote: ALZET pumps should only be filled with these specially designed filling tubes