Learn More
Sigma Aldrich Fine Chemicals Biosciences fillpourR Dialysis Tube fu
Catalog No. 502901319
Encompass
To use cut dialysis tubing to length and knot one end. Slip the open end onto the tube funnel and secure with included rubber band or Scienceware(R) silicone rubber loops. A captive snap on cover has a small writing area for easy identification. Steam autoclavable at 121 deg C (250 deg f). 10 per bag.Supports dialysis tubes on edge of vessel and allows easy filling.
Catalog No. 50-290-1319 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAF14848000110EA
To use cut dialysis tubing to length and knot one end. Slip the open end onto the tube funnel and secure with included rubber band or Scienceware(R) silicone rubber loops. A captive snap on cover has a small writing area for easy identification. Steam autoclavable at 121 deg C (250 deg f). 10 per bag.Supports dialysis tubes on edge of vessel and allows easy filling.