Learn More
Med Vet International Skin Barrier Applicator 3M Cavilon Advanced 80 To 90% Strength Hexamethyldisiloxane / Acrylic Tetrapolymer / 2-Octyl Cyanoacrylate Individual Packet Sterile, 5050, 20/Case
Catalog No. 502831892
Encompass_Preferred
The product’s superior barrier performance allows/enables damaged skin to heal The protective barrier prevents, stops and reverses the effects of IAD Due to its ability to attach /adhere to wet, weeping (denuded) and/or dry skin and its exceptional barrier properties, the product does a superior job of protecting skin from irritants Protective barrier reduces pain associated with IAD and IAD care Forms a transparent coating on the skin
Catalog No. 50-283-1892 Supplier Med Vet International Supplier No. 1045502CS
The product’s superior barrier performance allows/enables damaged skin to heal The protective barrier prevents, stops and reverses the effects of IAD Due to its ability to attach /adhere to wet, weeping (denuded) and/or dry skin and its exceptional barrier properties, the product does a superior job of protecting skin from irritants Protective barrier reduces pain associated with IAD and IAD care Forms a transparent coating on the skin