Learn More
Electron Microscopy Sciences Film Casting Device Set
SDP
Supplier:  Electron Microscopy Sciences 7130501
Encompass_Preferred
An all glass apparatus. It casts uniformly thin films of parlodion, formvar, or butvar directly onto 1x3 microscope slides. The film casting solution can be used repeatedly. A built-in fine-pressure-release valve helps control the speed of drainage. The thickness of the film is controlled by the concentration of the film solution and the rate of the drainage. The unit requires 100 mls of film casting solution to start.
The unit comes complete with:500 ml capacity flask with built-in valves; air-in and air-out, film casting cylinder with cover, and 75 cc atomizer.
Catalog No. 50-930-978