Learn More
Electron Microscopy Sciences Film Developing Tank 4x5", 9.5 x 15.3 x 14.9 cm
SDP
Catalog No. 5026771
Encompass_Preferred
Hard plastic tanks and floating lids.
I.D.: 9.5 x 15.3 x 14.9 cm
Use: six of 4x5" Hangers No. 4A
Catalog No. 50-267-71 Supplier Electron Microscopy Sciences Supplier No. 74343
Hard plastic tanks and floating lids.
I.D.: 9.5 x 15.3 x 14.9 cm
Use: six of 4x5" Hangers No. 4A