Learn More
Electron Microscopy Sciences Film Developing Tank 8x10", 20 x 27.8 x 26 cm
SDP
Catalog No. 5026770
Encompass_Preferred
Hard plastic tanks and floating lids.
I.D.: 20 x 27.8 x 26 cm
Use: Twelve of 8 x 10"
Catalog No. 50-267-70 Supplier Electron Microscopy Sciences Supplier No. 74347
Hard plastic tanks and floating lids.
I.D.: 20 x 27.8 x 26 cm
Use: Twelve of 8 x 10"