Learn More
Hach Company Film Forming Amine Kit
Supplier:  Hach Company 3630000
Encompass
Hach s Film Forming Amines kit allows you to test ODA OLA and OLDA based film forming amines products using Hach s DR3900 and DR6000 spectrophotometers Film Forming Amines are expensive and overdosing can lead to more than just higher costs Overdosing can lead to the difficulty in gunk ballsin in your system Under dosing can lead to corrosion of your systems Test for Film Forming Amines at lower concentrations with greater accuracy to ensure your systems are adequately protected costs are kept in check and overdosing risks are mitigated
Catalog No. 50-002-07084