Learn More
Electron Microscopy Sciences Film Marker
SDP
Catalog No. 5025904
Encompass_Preferred
This device when fitted to the 8X magnifier (68125) enables you to view and check the 35 mm roll film while marking the desired frame by depressing the puncher (located on the side of the device). The puncher punches out a V-shaped piece of film on the perforation to mark the frame. When fitted in the reverse position to the 8X magnifier, you are able to view and check the slides.
Catalog No. 50-259-04 Supplier Electron Microscopy Sciences Supplier No. 68126
This device when fitted to the 8X magnifier (68125) enables you to view and check the 35 mm roll film while marking the desired frame by depressing the puncher (located on the side of the device). The puncher punches out a V-shaped piece of film on the perforation to mark the frame. When fitted in the reverse position to the 8X magnifier, you are able to view and check the slides.