Learn More
Perkin Elmer US LLC Film Polarizer for the Double Polarizer Drive
Catalog No. 502104404
Encompass_Preferred
The Double Depolarizer Drive is compatible with the LAMBDA 650/750/850/950/1050 spectrometers. It requires, but does not include, one or two of the polarization elements listed below. : Film Polarizer (B2205022) Glan-Thompson Polarizer Crystal (B0505284) Glan-Taylor Polarizer Crystal (N1010520) Depolarizer Crystal (B2205021)
Catalog No. 50-210-4404 Supplier Perkin Elmer US LLC Supplier No. B2205022
The Double Depolarizer Drive is compatible with the LAMBDA 650/750/850/950/1050 spectrometers. It requires, but does not include, one or two of the polarization elements listed below. : Film Polarizer (B2205022) Glan-Thompson Polarizer Crystal (B0505284) Glan-Taylor Polarizer Crystal (N1010520) Depolarizer Crystal (B2205021)