Learn More
Grainger Film Roll 4 in W 250 ft L
Catalog No. 502767440
Encompass_Preferred
The Grainger Approved Film Roll is a 4 inch wide, 250 foot long stretch wrap film made from polyethylene. It has 80 gauge thickness and is sold in a single roll. The film provides holding force and load containment. It can be used for bundling, palletizing, and unitizing applications. This cost-effective stretch film is designed to cling to itself and the load for stabilization.
Catalog No. 50-276-7440 Supplier Grainger Supplier No. 16Y895
The Grainger Approved Film Roll is a 4 inch wide, 250 foot long stretch wrap film made from polyethylene. It has 80 gauge thickness and is sold in a single roll. The film provides holding force and load containment. It can be used for bundling, palletizing, and unitizing applications. This cost-effective stretch film is designed to cling to itself and the load for stabilization.