Learn More
Grainger Film Tape 1 in x 1 in Amber 2.5 mil PK50
Catalog No. 502767515
Encompass_Preferred
The TapeCase Film Tape is a brown, 2-inch wide, pressure-sensitive tape made of polypropylene film backing and rubber adhesive. It is designed for packaging, sealing, splicing, and general purpose use. This tape has moderate adhesion and is resistant to UV rays, moisture, and chemicals. It can be used indoors or outdoors and works well at temperatures ranging from -20°F to 200°F. The tape is available in multiple lengths, with a standard length of 60 yards. It meets the ASTM D6123 specification for packaging tape. The tape is manufactured in the USA by TapeCase.
Catalog No. 50-276-7515 Supplier Grainger Supplier No. 16U720
The TapeCase Film Tape is a brown, 2-inch wide, pressure-sensitive tape made of polypropylene film backing and rubber adhesive. It is designed for packaging, sealing, splicing, and general purpose use. This tape has moderate adhesion and is resistant to UV rays, moisture, and chemicals. It can be used indoors or outdoors and works well at temperatures ranging from -20°F to 200°F. The tape is available in multiple lengths, with a standard length of 60 yards. It meets the ASTM D6123 specification for packaging tape. The tape is manufactured in the USA by TapeCase.