Learn More
Grainger Film Tape 1inx1 in Amber 2.5 mil PK250
Catalog No. 502765038
Encompass_Preferred
The TapeCase Film Tape is a brown, 2" wide, 60 yard long, pressure-sensitive packaging tape made in the USA. It features a natural rubber adhesive and a 3.6 mil biaxially oriented polypropylene film backing. This tape is designed for box sealing, splicing, and general purpose applications. It adheres well to fiberboard, kraft paper, and other packaging materials. The tape has good holding power and resists aging, abrasion, moisture, and chemicals. It can be used for sealing, holding, reinforcing, packaging, and splicing applications. This is a versatile, heavy-duty packaging tape suitable for warehouse, shipping room, industrial, and home use.
Catalog No. 50-276-5038 Supplier Grainger Supplier No. 16U715
The TapeCase Film Tape is a brown, 2" wide, 60 yard long, pressure-sensitive packaging tape made in the USA. It features a natural rubber adhesive and a 3.6 mil biaxially oriented polypropylene film backing. This tape is designed for box sealing, splicing, and general purpose applications. It adheres well to fiberboard, kraft paper, and other packaging materials. The tape has good holding power and resists aging, abrasion, moisture, and chemicals. It can be used for sealing, holding, reinforcing, packaging, and splicing applications. This is a versatile, heavy-duty packaging tape suitable for warehouse, shipping room, industrial, and home use.