Learn More
Grainger TapeCase PTFE Silicone Slick Surface Tape, Gray, 2 in x 5 yd
Catalog No. 502757581
Encompass_Preferred
This pressure-sensitive tape is constructed with 5 mil skived PTFE film and a high-temperature silicone adhesive. It is designed for applications requiring a non-stick surface, low friction, and excellent electrical insulation. The tape provides reliable performance in both mechanical and electrical environments, featuring heat resistance up to 500 °F.
  • Features a 5 mil skived PTFE film backing with a silicone adhesive
  • Supports operation across a wide temperature range of -100 °F to +500 °F
  • Provides a non-stick, low-friction surface suitable for diverse material compatibility
  • Meets UL 510A requirements for flame retardancy and cold resistance
  • Complies with Mil-A-A59474 Type 1, Class 2 and PMC 7115 standards
Catalog No. 50-275-7581 Supplier Grainger Supplier No. 15D587
This pressure-sensitive tape is constructed with 5 mil skived PTFE film and a high-temperature silicone adhesive. It is designed for applications requiring a non-stick surface, low friction, and excellent electrical insulation. The tape provides reliable performance in both mechanical and electrical environments, featuring heat resistance up to 500 °F.
  • Features a 5 mil skived PTFE film backing with a silicone adhesive
  • Supports operation across a wide temperature range of -100 °F to +500 °F
  • Provides a non-stick, low-friction surface suitable for diverse material compatibility
  • Meets UL 510A requirements for flame retardancy and cold resistance
  • Complies with Mil-A-A59474 Type 1, Class 2 and PMC 7115 standards