Learn More
Grainger Film Tape 2 in x 72 yd Green 3.3 mil
Catalog No. 502751561
Encompass_Preferred
The TAPECASE Film Tape 2 in x 72 yd is a 2 inch wide by 72 yard long clear polypropylene film tape. It features a pressure sensitive acrylic adhesive and has a tensile strength of 22 lbs/in. The tape is designed for packaging, sealing, splicing, labeling, and other industrial uses. It can adhere to paper, plastic, glass, and metal surfaces. The operating temperature range is 32 to 150 degrees Fahrenheit.
Catalog No. 50-275-1561 Supplier Grainger Supplier No. 15C572
The TAPECASE Film Tape 2 in x 72 yd is a 2 inch wide by 72 yard long clear polypropylene film tape. It features a pressure sensitive acrylic adhesive and has a tensile strength of 22 lbs/in. The tape is designed for packaging, sealing, splicing, labeling, and other industrial uses. It can adhere to paper, plastic, glass, and metal surfaces. The operating temperature range is 32 to 150 degrees Fahrenheit.