Learn More
Grainger Film Tape 3/8inx3/8in Amber 2.5mil PK100
Catalog No. 502765035
Encompass_Preferred
The TapeCase Film Tape is a brown, pressure-sensitive, waterproof, polyethylene film tape. It has an acrylic adhesive and is designed for sealing, packaging, and general purpose use. This 2" x 60 yard roll of tape has a tensile strength of 22 lbs/in width. It adheres well to paper, plastic, glass, wood, and metal surfaces. The operating temperature range is -20°F to 160°F.
Catalog No. 50-276-5035 Supplier Grainger Supplier No. 16U717
The TapeCase Film Tape is a brown, pressure-sensitive, waterproof, polyethylene film tape. It has an acrylic adhesive and is designed for sealing, packaging, and general purpose use. This 2" x 60 yard roll of tape has a tensile strength of 22 lbs/in width. It adheres well to paper, plastic, glass, wood, and metal surfaces. The operating temperature range is -20°F to 160°F.