Learn More
Grainger Film Tape 4 in x 5 yd Amber 2.5 mil
Catalog No. 502752759
Encompass_Preferred
The Grainger 15C594 is a 4 inch wide by 5 yard long polyimide film tape with an amber color. This high temperature resistant tape is made in the USA and can withstand temperatures up to 500°F. The tape has an acrylic adhesive and comes in a convenient dispenser box for easy use. With its high dielectric strength, this tape is well-suited for applications like wire harnessing, banding, and insulation.
Catalog No. 50-275-2759 Supplier Grainger Supplier No. 15C594
The Grainger 15C594 is a 4 inch wide by 5 yard long polyimide film tape with an amber color. This high temperature resistant tape is made in the USA and can withstand temperatures up to 500°F. The tape has an acrylic adhesive and comes in a convenient dispenser box for easy use. With its high dielectric strength, this tape is well-suited for applications like wire harnessing, banding, and insulation.