Learn More
Grainger Film Tape 4 in x 72 yd Green 3.3 mil
Catalog No. 502753260
Encompass_Preferred
The Grainger 15C573 is a 4 inch wide polyethylene masking tape in green color. It is a general use tape for holding, sealing, splicing, masking and surface protection applications. This tape features a crepe paper backing and rubber adhesive. It can be used for both indoor and outdoor applications. The tape is UL listed and conforms to CID A-A-1684B. It comes in a standard length roll of 60 yards.
Catalog No. 50-275-3260 Supplier Grainger Supplier No. 15C573
The Grainger 15C573 is a 4 inch wide polyethylene masking tape in green color. It is a general use tape for holding, sealing, splicing, masking and surface protection applications. This tape features a crepe paper backing and rubber adhesive. It can be used for both indoor and outdoor applications. The tape is UL listed and conforms to CID A-A-1684B. It comes in a standard length roll of 60 yards.