Learn More
Microsolv Technology Corporation Film, Zone Free, Sealing for Micro-Sample Management System or 96 Well Plates. 70um polyethylene top layer and inert white layer / Adhesive Free Zones. 10/PK.
Catalog No. 502270497
Encompass_Preferred
Film has 3.12" x 6" x 6" outer dimensions. Supplied in a 4" x 7" bag.
This item is not returnable. View return policy
Catalog No. 50-227-0497 Supplier Microsolv Technology Corporation Supplier No. 95296ZFFPK
Film has 3.12" x 6" x 6" outer dimensions. Supplied in a 4" x 7" bag.