Learn More
Maine Molecular Quality Controls Inc FilmArray Pneumonia/Pneumoniaplus Control
SDP
Catalog No. NC1598294
Encompass
FilmArray Pneumonia/Pneumoniaplus Control is intended for use as an external positive and negative assayed quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of bacteria, viruses, and antimicrobial resistance genes. FilmArray Pneumonia/Pneumoniaplus Control is composed of synthetic RNA and DNA specifically designed for and intended to be used solely with the FilmArray Pneumonia Panel and FilmArray Pneumonia Panel plus assays. The FilmArray Pneumonia/Pneumoniaplus Control is comprised of 12 tubes, 6 tubes of positive control and 6 tubes of negative control, 120µL each. FilmArray Pneumonia/Pneumoniaplus POSITIVE contains synthetic RNA and DNA suspended in a non-infectious solution of buffers, preservatives and stabilizers. FilmArray Pneumonia/Pneumoniaplus NEGATIVE contains buffers and preservatives.
Catalog No. NC1598294 Supplier Maine Molecular Quality Controls Inc Supplier No. M340
FilmArray Pneumonia/Pneumoniaplus Control is intended for use as an external positive and negative assayed quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of bacteria, viruses, and antimicrobial resistance genes. FilmArray Pneumonia/Pneumoniaplus Control is composed of synthetic RNA and DNA specifically designed for and intended to be used solely with the FilmArray Pneumonia Panel and FilmArray Pneumonia Panel plus assays. The FilmArray Pneumonia/Pneumoniaplus Control is comprised of 12 tubes, 6 tubes of positive control and 6 tubes of negative control, 120µL each. FilmArray Pneumonia/Pneumoniaplus POSITIVE contains synthetic RNA and DNA suspended in a non-infectious solution of buffers, preservatives and stabilizers. FilmArray Pneumonia/Pneumoniaplus NEGATIVE contains buffers and preservatives.