Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Filming Amine Refill Kit, Methyl Orange
SDP

Catalog No. 13299564
Change view
Click to view available options
Quantity:
20/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-564 20/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-564 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Refill kit for the K-1001 FFA test kit. Contains 20 self-filling ampoules and 20 double point ampoules packaged securely.

  • Filming amine reacts with methyl orange to form a colored complex that is extracted into an immiscible organic solvent.
  • The intensity of the resulting yellow color is directly related to the concentration of 'filming amine' in the sample. Results are expressed as ppm (mg/L) octadecylamine (ODA).

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-1001
No. of Tests 20
Quantity 20/Box
Type Filming Amine Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.