Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Filming Amine Refill Kit, Rose Bengal
SDP

Catalog No. 13299701
Change view
Click to view available options
Quantity:
30/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-701 30/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-701 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R1006
Add to Cart
Add to Cart

Refill kit for the K-1006 FFA test kit. Contains 30 self-filling ampoules packaged securely in trays.

  • When samples are buffered under acidic conditions (pH 2.3 - 3.3), filming amines such as Octadecylamine (ODA) and Oleylamine (OLA) form a magenta-colored complex with rose bengal (4,5,6,7-tetrachloro-2',4',5',7'-tetraiodofluorescein disodium salt) in direct proportion to the concentration of filming amines in the sample.
  • Results are expressed as ppm (mg/L) ODA or OLA.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-1006
No. of Tests 30
Quantity 30/Box
Type Filming Amine Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.