Learn More
Premier Lab Supply FilmVelopes, PN# FV-135-212 – Mylar film 3.5μm (0.14mil)/2.0” Diameter
SDP
Catalog No. NC1547115
Encompass
FilmVelopes - CLEAN, EASY & FASTER sample prep without the tear away! (PN# FV-135-212). Innovative proprietary thin film applicator utilizing specialty jackets firmly holding a thin film in place while allowing for a clean release upon application. FilmVelopes reduces the probability of error and excess film waste while allowing for easy handling. 100 per pack.
Catalog No. NC1547115 Supplier Premier Lab Supply Supplier No. FV135212
FilmVelopes - CLEAN, EASY & FASTER sample prep without the tear away! (PN# FV-135-212). Innovative proprietary thin film applicator utilizing specialty jackets firmly holding a thin film in place while allowing for a clean release upon application. FilmVelopes reduces the probability of error and excess film waste while allowing for easy handling. 100 per pack.