Learn More
Perkin Elmer US LLC PTFE Filter for FIMS Gas-Liquid Separator, Pkg. 5
Catalog No. 509052026
Encompass_Preferred
PTFE Filter Disks for the Gas-Liquid Separator of the Flow Injection Mercury System (FIMS). For a successful analysis, the filter disk of the gas-liquid separator must be inspected on a regular basis since it is required that it is clean and dry. The disks are sold in packages of 5.
Catalog No. 50-905-2026 Supplier Perkin Elmer US LLC Supplier No. B0508567
PTFE Filter Disks for the Gas-Liquid Separator of the Flow Injection Mercury System (FIMS). For a successful analysis, the filter disk of the gas-liquid separator must be inspected on a regular basis since it is required that it is clean and dry. The disks are sold in packages of 5.