Learn More
Cole-Parmer Argos Technologies Syringe Filter, 13mm, .22um, Blue PP/PVDF, Sterile; 100/Pk
Supplier:  Cole-Parmer 0439632
Encompass_Preferred
Minimize sample loss. Choose the membrane material best suited for your application. Each filter is quality tested for filter efficiency and housing integrity for use up to 75 psi (5.1 bar). Sterile filters are certified nonpyrogenic, have been gamma sterilized, and come individually packaged with lot numbers printed on for easy traceability.
Catalog No. 50-196-1410